Anti-CD46 Mouse Monoclonal Antibody [clone: 122.2]

Toimittaja: ANTIBODIES.COM
A249308-100
ANTIA249308-100EA 520 EUR
ANTIA249308-100
Anti-CD46 Mouse Monoclonal Antibody [clone: 122.2]
Vasta-aineet
Mouse monoclonal [122.2] antibody to CD46 for functional studies, flow cytometry and IF with samples derived from human.

  • Validated applications: Functional studies, Flow cytometry, IF
  • Recommended dilutions: Flow cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml

Type: Primary
Antigen: CD46
Clonality: Monoclonal
Clone: 122.2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR