Anti-Cadherin 16 Mouse Monoclonal Antibody [clone: SPM594]

Toimittaja: ANTIBODIES.COM
A248066-100
ANTIA248066-100EA 520 EUR
ANTIA248066-100
Anti-Cadherin 16 Mouse Monoclonal Antibody [clone: SPM594]
Vasta-aineet
Mouse monoclonal [SPM594] antibody to Cadherin 16 for WB and IHC-P with samples derived from human, mouse, rat, rabbit and canine.

  • Validated applications: WB, IHC-P
  • Recommended dilutions: WB: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: Cadherin 16
Clonality: Monoclonal
Clone: SPM594
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse, Rat, Rabbit, Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR