Anti-CD326 Mouse Monoclonal Antibody [clone: 323/A3] (FITC (Fluorescein Isothiocyanate))

Toimittaja: ANTIBODIES.COM
A122123-100
ANTIA122123-100EA 300 EUR
ANTIA122123-100
Anti-CD326 Mouse Monoclonal Antibody [clone: 323/A3] (FITC (Fluorescein Isothiocyanate))
Vasta-aineet
Mouse monoclonal [323/A3] antibody to CD326 (FITC) for Flow Cytometry with samples derived from human.

  • Validated applications: Flow cytometry
  • Recommended dilutions: Flow cytometry: This reagent is designed for analysis of human blood cells using 4 µl reagent / 100 µl of whole blood or 106 cells in a suspension.

Type: Primary
Antigen: CD326
Clonality: Monoclonal
Clone: 323/A3
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR