Anti-C Reactive Protein Rabbit Polyclonal Antibody

Toimittaja: ANTIBODIES.COM
A100056-100
ANTIA100056-100EA 475 EUR
ANTIA100056-100
Anti-C Reactive Protein Rabbit Polyclonal Antibody
Vasta-aineet
Rabbit polyclonal antibody to CRP for WB and ELISA with samples derived from human and rat.

  • Validated Applications: WB, ELISA
  • Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000

Type: Primary
Antigen: C Reactive Protein
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR