Chemical resistant overalls, AlphaTec® 1800 COMFORT, Model 195
Toimittaja: Ansell
WN18B-00195-02 WN18B-00195-07 WN18B-00195-03 WN18B-00195-08 WN18B-00195-09 WN18B-00195-06 WN18B-00195-05 WN18B-00195-04
ANSEWN18B-00195-02EA 317 EUR
ANSEWN18B-00195-02 ANSEWN18B-00195-07 ANSEWN18B-00195-03 ANSEWN18B-00195-08 ANSEWN18B-00195-09 ANSEWN18B-00195-06 ANSEWN18B-00195-05 ANSEWN18B-00195-04
Chemical resistant overalls, AlphaTec® 1800 COMFORT, Model 195
Kertakäyttöiset vaatteet
Lightweight, breathable microporous laminate technology, offering Type 5/6 protection.
  • Protects against low hazard liquid spray and fine particulates
  • Comfortable to wear featuring moisture vapor permeable fabric to help reduce the risk of heat stress
  • Optimized body fit, improving wearer comfort and safety
  • Anti-static, tested according to EN 1149-5 and AATCC 76
  • Low linting to help reduce the risk of cross contamination in critical areas
  • Silicone free, critical in spray painting applications
Designed for workers in warm environments to provide anMore Product Information