Chemical resistant overalls, AlphaTec® 5000, Model 125-G02
Toimittaja: Ansell
OR50T-12502G02 OR50T-12508G02 OR50T-12504G02 OR50T-12505G02 OR50T-12507G02 OR50T-12509G02 OR50T-12506G02 OR50T-12503G02
ANSEOR50T-12502G02EA 1400 EUR
ANSEOR50T-12502G02 ANSEOR50T-12508G02 ANSEOR50T-12504G02 ANSEOR50T-12505G02 ANSEOR50T-12507G02 ANSEOR50T-12509G02 ANSEOR50T-12506G02 ANSEOR50T-12503G02
Chemical resistant overalls, AlphaTec® 5000, Model 125-G02
Kertakäyttöiset vaatteet
Durable, highly visible, multi-layer chemical barrier technology offering Type 3/4/5 protection.
  • Outstanding permeation performance to organic and inorganic chemicals, biological hazards and chemical warfare agents
  • Exceptional barrier protection, providing >480 minutes breakthrough time against 14 of 15 chemicals listed in EN ISO 6529
  • Highly visible bright orange colour for improved worker safety
  • Welded and taped seams, offering the highest barrier to liquids and particulates
  • Multi-layer barrier material is lightweight, yet strong and durable
  • Anti-