General purpose hydrometers, S50
Toimittaja: Alla France
3502SC07060BQP 3502SC08060BQP 3502SC08560BQP 3502SC09060BQP 3502SC10560MQP 3502SC11060MQP 3502SC12060MQP 3502SC12560MQP 3502SC19560HQP 3502SC16060HQP 3502SC16560HQP 3502SC17060HQP 3502SC17560HQP 3502SC18060HQP 3502SC18560HQP 3502SC19060HQP
ALLA3502SC07060BQPEA 55.6 EUR
ALLA3502SC07060BQP ALLA3502SC08060BQP ALLA3502SC08560BQP ALLA3502SC09060BQP ALLA3502SC10560MQP ALLA3502SC11060MQP ALLA3502SC12060MQP ALLA3502SC12560MQP ALLA3502SC19560HQP ALLA3502SC16060HQP ALLA3502SC16560HQP ALLA3502SC17060HQP ALLA3502SC17560HQP ALLA3502SC18060HQP ALLA3502SC18560HQP ALLA3502SC19060HQP
General purpose hydrometers, S50
Ominaispainomittarit
General purpose hydrometers graduated in SG and calibrated at 60 °F (15 °C).
Sertifikaatit: ISO 650