Bravo NGS
Toimittaja: AGILENT
G5573AA
AGLSG5573AAEA 203645 EUR
AGLSG5573AA
Bravo NGS
Sample Preparation Instruments
Automate genomics workflows for NGS without sacrificing quality, in a small footprint.
  • Quickly automate with ready-to-use protocols and pre-developed methods for Agilent SureSelect, Agilent HaloPlex, Illumina TruSeq, NuGEN Ovation, KAPA HTP, PacBio SMRTbell, NEBNext, and more
  • Easy to use interface requires no programming expertise to run your automated methods
  • Consistent data quality allows you to increase your throughput, with consistent results every time
  • Prepare up to ten times more samples in the same time
  • Open platform allows you to automate more NGS