DNase I

Toimittaja: AGILENT

600031 600032
AGLS600031EA 197 EUR
AGLS600031 AGLS600032
DNase I
Entsyymit

Purified DNase I tested to be free of contaminating activity for the degradation of nucleic acids.


  • RNase free
  • Contains a mixture of four isoenzymes from bovine pancreas (10 units/µl)


DNase I, RNase Free catalyses the degradation of double-stranded DNA into oligonucleotides 1,2 and mononucleotides. This enzyme has been isolated as a mixture of four isoenzymes from bovine pancreas that cut preferentially next to pyrimidine nucleotides.


Reaction Conditions: 40 mM Tris-HCl (pH 7,5), 6 mM MgCl₂, 2 mM CaCl₂, 1 µg DNA and 1 unit enzyme in a 25 µl volume. Incubate 30 minutes at 37 °C.


Varoitus: For research use only. Not for use in diagnostic procedures.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR