Multiple affinity removal spin cartridge reagent kit
Toimittaja: AGILENT
5188-5249 5188-5250 5188-5251 5188-5252 5188-5254
AGLS5188-5249EA 9 EUR
AGLS5188-5249 AGLS5188-5250 AGLS5188-5251 AGLS5188-5252 AGLS5188-5254
Multiple affinity removal spin cartridge reagent kit
Proteiininpuhdistusjärjestelmät Protein Purification Kits
The affinity removal spin cartridge reagent kit is designed to remove the most interfering high-abundant protein (albumin) from human samples (such as plasma, serum, urine or cerebrospinal fluid).
Toimitustiedot: Kit includes Buffer A, 1 bottle (1 L) Buffer B, 1 bottle (1 L), Spin filters, 2 packs, Spin concentrators, 5 KDa MWCo (4 ml) 1 pack, Luer-Lok adapter 1 pack, Luer-Lok syringes (5 ml) 1 pack, Microtubes, 6 packs, Caps and plugs 1 pack, PTFE Luer-Lok needles 1 pack.