Multiple affinity removal starter reagent kits
Toimittaja: AGILENT
5185-5986 5185-5987 5185-5988 5185-5991
AGLS5185-5986EA 770 EUR
AGLS5185-5986 AGLS5185-5987 AGLS5185-5988 AGLS5185-5991
Multiple affinity removal starter reagent kits
Proteiininpuhdistusjärjestelmät Protein Purification Kits
Starter reagent kit for multiple affinity removal LC columns.
Toimitustiedot: Includes: 2 bottles buffer A (1 L), 1 bottle buffer B (1 L), 2 packs of 25 spin filters, 1 pack of 25 spin concentrator.