High Sensitivity RNA ScreenTape Analysis
Toimittaja: AGILENT
5067-5579 5067-5580 5067-5581
AGLS5067-5579EA 249 EUR
AGLS5067-5579 AGLS5067-5580 AGLS5067-5581
High Sensitivity RNA ScreenTape Analysis
Elektroforeesi Kapillaarielektroforeesijärjestelmät
Reliable, reproducible, and objective measurements of RNA samples of limited abundance.
  • Utilises RINe – a user independent algorithm for accurate and objective assessment of eukaryotic or prokaryotic total RNA integrity
  • Fast results are obtained in approximately 2 minutes per sample
  • Minimal sample consumption is required as only 2 µl of sample is needed
  • Detection of RNA sample with limited abundance down to 100 pg/µl is possible
  • Objective quality control of precious RNA samples is enabled within RNAseq, microarray analysis, and quantitative RT-PCR