Bioanalyzer High Sensitivity Protein Analysis
Toimittaja: AGILENT
5067-1575 5067-1576 5067-1577 5067-1578
AGLS5067-1575EA 1150 EUR
AGLS5067-1575 AGLS5067-1576 AGLS5067-1577 AGLS5067-1578
Bioanalyzer High Sensitivity Protein Analysis
Elektroforeesi Kapillaarielektroforeesijärjestelmät
High sensitivity electrophoretic separation of proteins and peptides up to 250 kD.
  • A ready-to-use system featuring standardised assays and pre-packaged reagent kits which are suitable to analyse samples up to 250 kD
  • After sample labelling, fast results are achieved with automated electrophoresis of 10 samples in 30 minutes
  • Highly sensitive with an accuracy down to silver stain levels, approximately 1 pg/µl of labelled protein on chip
  • Fully operational across a large linear dynamic range with four orders of magnitude
  • Highly precise impurity detection, down