PCR and qPCR consumables
Toimittaja: AGILENT HEWLETT PACKARD
410094 401333 401334
HEWL410094EA 0 EUR
HEWL410094 AGLS401333 AGLS401334
PCR and qPCR consumables
PCR-laitteet
High-quality well plate supplies and accessories will enhance your PCR and qPCR workflow and analysis.
Agilent AriaMx low profile strip tubes are optimised for PCR and qPCR applications. The tubes are validated for optimal performance with Mx3005P, Mx3000P, and the AriaMx real time PCR Systems. Part of the qPCR Plastics and Accessories, the products include the perfect fit frame, tubes, and caps.
Additional products available include the 96-well semiskirted polypropylene PCR plate compatible with the optical Strip Caps, the StrataCoolers LP Benchtop Cooler, and StrataCooler cryoMore Product Information