Reservoir Microplates
Toimittaja: AGILENT
201254-100 201244-100 200686-100 204363-100 204484-100 201250-100 204093-100 201252-100 204504-100 204017-100 201246-100 201256-100 204095-100 201280-100 204365-100 201284-100 204284-100
AGLS201254-100EA 152 EUR
AGLS201254-100 AGLS201244-100 AGLS200686-100 AGLS204363-100 AGLS204484-100 AGLS201250-100 AGLS204093-100 AGLS201252-100 AGLS204504-100 AGLS204017-100 AGLS201246-100 AGLS201256-100 AGLS204095-100 AGLS201280-100 AGLS204365-100 AGLS201284-100 AGLS204284-100
Reservoir Microplates
Kuoppalevyt Levyt suurtehoseulontaan
Polypropylene reservoir microplates for automated liquid handlers and handheld pipettors.
  • Large range of cavity volumes, from 2,6 to 300 ml with specialised bottom geometries to optimise access and minimise dead volumes
  • Low binding surfaces for minimal losses of proteins, DNA/RNA, and other biologicals
  • Molded from chemical-resistant polymers, for maintained performance following prolonged exposure to aggressive solutions and solvents
  • Produced to ANSI/SLAS standards, for compatibility with popular labware and devices
  • Compatible with all major platforms and