JM competent cells
Toimittaja: AGILENT
200234 200235 200239
AGLS200234EA 115 EUR
AGLS200234 AGLS200235 AGLS200239
JM competent cells
Kompetentit solut
JM Competent Cells are recombination- and E. coli K12 restriction-proficient.
  • Blue-white color screening possible for recombinant plasmids
  • Transformation efficiency >1×10⁸ transformants/µg DNA
  • High quality competent cells for preparing methylation-free DNA
  • The SCS110 strain lacks both dam and dcm activity, and DNA propagated in this strain can be digested by methylation-sensitive enzymes such as Xba I, Cla I and EcoR II
  • JM110 cells contain the lacIqZΔM15 gene on the F' episome, allowing blue-white screening for recombinant plasmids