System Calibration Standard (7 components)
Toimittaja: CPACHEM
F237594
ACSDF237594EA 509 EUR
ACSDF237594
System Calibration Standard (7 components)
Standardiaineet ja vertailumateriaalit Petroleum Industry Standards
System calibration standard with 7 components.
  • Solvent: n-Heptane
  • Volume: ampoule 5 ml
The components consists of cyclohexane 1% (w/v), 1-phenyldodecane 0,1% (w/v), o-Xylene (1,2-dimethylbenzene) 0.5% (w/v), hexamethylbenzene 0,1% (w/v), naphthalene 0,1% (w/v), dibenzothiophene 0,05% (w/v) 9-methylanthracene 0,05% (w/v).
Sertifikaatit: EN 12916