Dexmedetomidine hydrochloride

Toimittaja: Thermo Fisher Scientific
Varoitus

465430010 465432500
ACRO465430010EA 694 EUR
ACRO465430010 ACRO465432500
Dexmedetomidine hydrochloride
Dexmedetomidine hydrochloride

  • Bioactive

Kaava: C₁₃H₁₆N₂·HCl
Sulamispiste: 158 °C
Storage Temperature: Jääkaappi
MDL numero: MFCD12912677
CAS-numero: 145108-58-3

Order Now

Testitulosten erittely

Appearance White to beige Powder
Infrared spectrum Conforms
Melting point 156 to 160 °C
HPLC ≥97.5 %
Specific optical rotation +48° to +54° (20 °C, 589 nm) (1% in water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR