Oxybutynin hydrochloride

Toimittaja: Thermo Fisher Scientific
Varoitus

463500010 463500050 463500250
ACRO463500010EA 118 EUR
ACRO463500010 ACRO463500050 ACRO463500250
Oxybutynin hydrochloride
Oxybutynin hydrochloride

  • Bioactive

Kaava: C₂₂H₃₁NO₃·HCl
Sulamispiste: 125…128 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00072150
CAS-numero: 1508-65-2

Order Now

Testitulosten erittely

Appearance White to almost white Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤3 %
Titration with NaOH 97.5 to 102.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR