Caffeine citrate

Toimittaja: Thermo Fisher Scientific
Varoitus

Synonyms: 2-hydroxypropane-1,2,3-tricarboxylic acid, citric acid,1,3,7-trimethylpurine-2,6-dione, Citrated caffeine

461720250 461721000
ACRO461720250EA 128 EUR
ACRO461720250 ACRO461721000
Caffeine citrate
Caffeine citrate
Kaava: C₁₄H₁₈N₄O₉
Sulamispiste: 159…161 °C
Storage Temperature: Jääkaappi
MDL numero: MFCD00044986
CAS-numero: 69-22-7

Order Now

Testitulosten erittely

Appearance White to off-white Powder
Infrared spectrum Conforms
Loss on drying ≤5 %
Titration with HClO4 48 to 52 % (Caffeine)
Titration with TBAH 48 to 52 % (Citric acid)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR