Gliclazide

Toimittaja: Thermo Fisher Scientific
Varoitus

461070010 461070050
ACRO461070010EA 49.5 EUR
ACRO461070010 ACRO461070050
Gliclazide
Gliclazide
Kaava: C₁₅H₂₁N₃O₃S
Sulamispiste: 163…169 °C
Storage Temperature: Jääkaappi
MDL numero: MFCD00409893
CAS-numero: 21187-98-4

Order Now

Testitulosten erittely

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 163 to 169 °C
Loss on drying ≤0.5 % (1 g, 105 °C)
HPLC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR