Gemcitabine hydrochloride 98%

Toimittaja: Thermo Fisher Scientific
Vaara

461060010 461060050
ACRO461060010EA 127 EUR
ACRO461060010 ACRO461060050
Gemcitabine hydrochloride 98%
Gemcitabine hydrochloride
Kaava: C₉H₁₂ClF₂N₃O₄
Storage Temperature: Jääkaappi
MDL numero: MFCD01735988
CAS-numero: 122111-03-9

Order Now

Testitulosten erittely

Appearance White to off-white Powder
Infrared spectrum Conforms
Titration Argentometric 98 to 102 %
HPLC ≥97.5 %
Optical rotation +43° to +50° (1 % in water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR