Naftifine hydrochloride 99%

Toimittaja: Thermo Fisher Scientific

458660010 458660050 458660250
ACRO458660010EA 77.5 EUR
ACRO458660010 ACRO458660050 ACRO458660250
Naftifine hydrochloride 99%
Naftifine hydrochloride
Kaava: C₂₁H₂₁N
Sulamispiste: 175…179 °C
Storage Temperature: Jääkaappi
MDL numero: MFCD00059047
CAS-numero: 65473-14-5

Order Now

Testitulosten erittely

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 174 to 180 °C
Titration with HClO4 ≥98.5 %
Loss on fusion ≤0.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR