Lithium tetrakis(pentafluorophenyl)borate ethyl etherate

Toimittaja: Thermo Fisher Scientific
Varoitus

458530010 458531000
ACRO458530010EA 307 EUR
ACRO458530010 ACRO458531000
Lithium tetrakis(pentafluorophenyl)borate ethyl etherate
Lithium tetrakis(pentafluorophenyl)borate ethyl etherate
Kaava: C₂₄BF₂₀Li·2.5C₄H₁₀O
Sulamispiste: 117…122 °C
Storage Temperature: Huoneenlämpö
CAS-numero: 371162-53-7

Order Now

Testitulosten erittely

Appearance White to light beige Powder or crystals
Melting point 117 to 122°C
NMR (19F-NMR) ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR