Chloroquine diphosphate salt 98%

Toimittaja: Thermo Fisher Scientific
Varoitus

455240250 455241000 455245000
ACRO455240250EA 60.3 EUR
ACRO455240250 ACRO455241000 ACRO455245000
Chloroquine diphosphate salt 98%
Chloroquine diphosphate salt
Kaava: C₁₈H₂₆ClN₃
Sulamispiste: 200 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00069852
CAS-numero: 50-63-5

Order Now

Testitulosten erittely

Appearance White to beige Powder
Infrared spectrum Conforms
Loss on drying ≤2 % (105°C, 16 h)
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR