Kaliumnitriitti 97%, extra pure

Toimittaja: Thermo Fisher Scientific
Vaara

453190010 453190050
ACRO453190010EA 103 EUR
ACRO453190010 ACRO453190050
Kaliumnitriitti 97%, extra pure
Kaliumnitriitti
Kaava: KNO₂
Sulamispiste: 387 °C
Tiheys: 1,92 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011408
CAS-numero: 7758-09-0
UN: 1488
ADR: 5.1,II

Order Now

Testitulosten erittely

Appearance White to light yellow Powder or crystals
Titration excess KMnO4 96.0 to 104.0 %
Arsenic (As) ≤1 ppm
Heavy metals (as Pb) ≤10 ppm
Insoluble matter ≤0.01 % (10 % in water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR