Chloromethylphosphonic acid 98%

Toimittaja: Thermo Fisher Scientific
Vaara

451600010 451600050
ACRO451600010EA 113 EUR
ACRO451600010 ACRO451600050
Chloromethylphosphonic acid 98%
Chloromethylphosphonic acid
Kaava: CH₄ClO₃P
Sulamispiste: 88…92 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00013955
CAS-numero: 2565-58-4
UN: 3261
ADR: 8,II

Order Now

Testitulosten erittely

Appearance White to brown Crystalline powder
Infrared spectrum Conforms
Melting point 88 to 92 °C
Titration with NaOH ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR