Diphenyliodonium hexafluorophosphate 97%

Toimittaja: Thermo Fisher Scientific
Vaara

450910050 450910250 450911000
ACRO450910050EA 58.1 EUR
ACRO450910050 ACRO450910250 ACRO450911000
Diphenyliodonium hexafluorophosphate 97%
Diphenyliodonium hexafluorophosphate
Kaava: C₁₂H₁₀F₆IP
Sulamispiste: 140…144 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00061398
CAS-numero: 58109-40-3
UN: 1759
ADR: 8,II

Order Now

Testitulosten erittely

Appearance White to off-white Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 138 to 143 °C
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR