Trinatriumfosfaatti-12 H2O, extra pure

Toimittaja: Thermo Fisher Scientific
Varoitus

450660010 450660050 450660250
ACRO450660010EA 40.2 EUR
ACRO450660010 ACRO450660050 ACRO450660250
Trinatriumfosfaatti-12 H2O, extra pure
Trinatriumfosfaatti-12 H2O
Kaava: Na₃PO₄·12H₂O
Sulamispiste: 75 °C
Tiheys: 1,62 g/cm³
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00149198
CAS-numero: 10101-89-0

Order Now

Testitulosten erittely

Appearance Colorless to white Powder or crystals or needles
Titration excess HCl ≥97.5 %
Free alkali (NaOH) ≤3.5 %
Insoluble matter ≤0.05 % (in water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR