5-Acetylvaleric acid 90+%, Technical Grade

Toimittaja: Thermo Fisher Scientific
Vaara

449530050 449530250 449531000
ACRO449530050EA 55.2 EUR
ACRO449530050 ACRO449530250 ACRO449531000
5-Acetylvaleric acid 90+%, Technical Grade
5-Acetylvaleric acid
Kaava: C₇H₁₂O₃
Kiehumispiste: 138…140 °C (1,5 mmHg)
Sulamispiste: 35…37 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00051479
CAS-numero: 3128-07-2
UN: 3261
ADR: 8,II

Order Now

Testitulosten erittely

Appearance White to off-white Powder or crystals
Infrared spectrum Conforms
GC ≥90.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR