Cortisone acetate 98+%

Toimittaja: Thermo Fisher Scientific
Varoitus

448960050 448960250
ACRO448960050EA 133 EUR
ACRO448960050 ACRO448960250
Cortisone acetate 98+%
Cortisone acetate
Kaava: C₂₃H₃₀O₆
Sulamispiste: 237…240 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00003609
CAS-numero: 50-04-4

Order Now

Testitulosten erittely

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤0.5 %
HPLC ≥98.0 %
Specific optical rotation +207° to +220° (20°C, 589 nm) (c=1, dioxane)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR