Magnesiumkloridihexahydraatii 98+%, extra pure

Toimittaja: Thermo Fisher Scientific

447700010 447700050
ACRO447700010EA 48.5 EUR
ACRO447700010 ACRO447700050
Magnesiumkloridihexahydraatii 98+%, extra pure
Magnesiumkloridihexahydraatii
Kaava: MgCl₂·6H₂O
Kiehumispiste: 1412 °C (1013 hPa)
Sulamispiste: 117 °C
Tiheys: 1,57 g/cm³ (25 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00149781
CAS-numero: 7791-18-6

Order Now

Testitulosten erittely

Appearance White Crystalline powder
Titration Complexometric 98.0 to 101.0 %
Water 51.0 to 55.0 %
Heavy metals (as Pb) ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR