4,4,4,4',4',4'-Hexafluoro-DL-valine 97%

Toimittaja: Thermo Fisher Scientific
Varoitus

446540010 446540050
ACRO446540010EA 234 EUR
ACRO446540010 ACRO446540050
4,4,4,4',4',4'-Hexafluoro-DL-valine 97%
4,4,4,4',4',4'-Hexafluoro-DL-valine
Kaava: C₅H₅F₆NO₂
Sulamispiste: 207…211 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00014349
CAS-numero: 16063-80-2

Order Now

Testitulosten erittely

Appearance White to off-white Powder or solid
Infrared spectrum Conforms
Assay 96 to 104 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR