Tetrahydrofuraani 99.5% stabilised for analysis

Toimittaja: Thermo Fisher Scientific
Vaara

Synonyms: Butylene oxide, THF, Diethylene oxide

Stabilization: Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)

446400010 446400025
ACRO446400010EA 65.9 EUR
ACRO446400010 ACRO446400025
Tetrahydrofuraani 99.5% stabilised for analysis
Tetrahydrofuraani
Kaava: C₄H₈O
Kiehumispiste: 66 °C (760 mmHg)
Sulamispiste: –108,4 °C
Tiheys: 0,88 g/cm³
Leimahduspiste: –21 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00005356
CAS-numero: 109-99-9
UN: 2056
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b2d7611caf0127fbef941994edd1d7e69487d54a&vid=2a64e21bd2e13636c3038adf839b3e9ff18509c5&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4060 to 1.4080 (20°C, 589 nm)
Water ≤0.05 %
GC ≥99.4 %
Color scale ≤10 APHA
Stabilizer 0.025% BHT
Cadmium (Cd) ≤0.05 ppm
Calcium (Ca) ≤0.5 ppm
Chromium (Cr) ≤0.05 ppm
Cobalt (Co) ≤0.05 ppm
Copper (Cu) ≤0.05 ppm
Iron (Fe) ≤0.5 ppm
Lead (Pb) ≤0.1 ppm
Magnesium (Mg) ≤0.5 ppm
Manganese (Mn) ≤0.05 ppm
Nickel (Ni) ≤0.02 ppm
Zinc (Zn) ≤0.5 ppm
Acidity (CH3COOH) ≤0.001 %
Peroxides ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR