4-Thiazolecarboxylic acid 97%

Toimittaja: Thermo Fisher Scientific
Varoitus

443970010 443970050
ACRO443970010EA 75.8 EUR
ACRO443970010 ACRO443970050
4-Thiazolecarboxylic acid 97%
4-Thiazolecarboxylic acid
Kaava: C₄H₃NO₂S
Sulamispiste: 195…199 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00623589
CAS-numero: 3973-08-8

Order Now

Testitulosten erittely

Appearance White to yellow to beige Crystalline powder
Infrared spectrum Conforms
Melting point 190 to 205 °C
Loss on drying ≤3 %
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR