Cyclopropylacetic acid 97%

Toimittaja: Thermo Fisher Scientific
Vaara

443780010 443780050
ACRO443780010EA 84.5 EUR
ACRO443780010 ACRO443780050
Cyclopropylacetic acid 97%
Cyclopropylacetic acid
Kaava: C₅H₈O₂
MW: 100.12 g/mol
Kiehumispiste: 60…61 °C (29 mmHg)
Tiheys: 1.14 g/cm³
Leimahduspiste: 93 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00041544
CAS-numero: 5239-82-7
UN: 3265
ADR: 8 II

Order Now

Testitulosten erittely

Appearance Colorless to light yellow Liquid
Infrared spectrum Conforms
Titration with NaOH ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR