tert-Butyl-3-oxoazetidine-1-carboxylate 97%

Toimittaja: Thermo Fisher Scientific
Varoitus

442040010 442040050
ACRO442040010EA 90.9 EUR
ACRO442040010 ACRO442040050
tert-Butyl-3-oxoazetidine-1-carboxylate 97%
tert-Butyl-3-oxoazetidine-1-carboxylate
Kaava: C₈H₁₃NO₃
Sulamispiste: 50…52 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD01861741
CAS-numero: 398489-26-4
UN: 3335
ADR: 9,III

Order Now

Testitulosten erittely

Appearance White to light brown Crystalline powder or solid
Infrared spectrum Conforms
Melting point 48 to 54 °C
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR