2-Amino-4-chloropyridine 97%

Toimittaja: Thermo Fisher Scientific
Varoitus

Synonyms: 4-Chloro-2-pyridylamine, 4-Chloropyridin-2-amine

441460050 441460250
ACRO441460050EA 67.6 EUR
ACRO441460050 ACRO441460250
2-Amino-4-chloropyridine 97%
2-Amino-4-chloropyridine
Kaava: C₅H₅ClN₂
MW: 128.56 g/mol
Sulamispiste: 130…132 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD04113820
CAS-numero: 19798-80-2

Order Now

Testitulosten erittely

Appearance White to light brown Solid
Infrared spectrum Conforms
Melting point 127 to 134 °C
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR