D-Biotin-N-succinimidyl ester 98%

Toimittaja: Thermo Fisher Scientific

439302500 439300010
ACRO439302500EA 262 EUR
ACRO439302500 ACRO439300010
D-Biotin-N-succinimidyl ester 98%
D-Biotin-N-succinimidyl ester
Kaava: C₁₄H₁₉N₃O₅S
Sulamispiste: 202,3…202.7 °C
Storage Temperature: Pakastin
MDL numero: MFCD00078531
CAS-numero: 35013-72-0

Order Now

Testitulosten erittely

Appearance White to off-white Powder
Infrared spectrum Conforms
Water ≤2 %
Elemental Analysis 12.00 to 12.62 % N
Specific optical rotation +52° to +54° (20°C, 589 nm) (c=1 in DMF)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR