Cyclopentanesulphonyl chloride 90%

Toimittaja: Thermo Fisher Scientific
Vaara

437830010 437830050
ACRO437830010EA 173 EUR
ACRO437830010 ACRO437830050
Cyclopentanesulphonyl chloride 90%
Cyclopentanesulphonyl chloride
Kaava: C₅H₉ClO₂S
Kiehumispiste: 95…98 °C (2Torr)
Leimahduspiste: >110 °C
Storage Temperature: Jääkaappi
MDL numero: MFCD04117774
CAS-numero: 26394-17-2
ADR: 8,II

Order Now

Testitulosten erittely

Appearance Light brown to dark brown Liquid
Infrared spectrum Conforms
GC ≥88.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR