Indium(III) acetate, vedetön 99.99% (trace metals basis)

Toimittaja: Thermo Fisher Scientific
Varoitus

436980100 436980500
ACRO436980100EA 107 EUR
ACRO436980100 ACRO436980500
Indium(III) acetate, vedetön 99.99% (trace metals basis)
Indium(III) acetate
Kaava: C₂H₄InO₂
Sulamispiste: 270 °C (haj. lämpötila)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00054347
CAS-numero: 25114-58-3

Order Now

Testitulosten erittely

Appearance White Crystalline powder
Titration Complexometric 35.3 to 43.3 % (In)
Total trace metal impurities ≤100 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR