Nikkeli 99.995% Trace metals basis, powder

Toimittaja: Thermo Fisher Scientific
Vaara

436780500
ACRO436780500EA 116 EUR
ACRO436780500
Nikkeli 99.995% Trace metals basis, powder
Nikkeli
Kaava: Ni
MW: 58.69 g/mol
Kiehumispiste: 3007
Sulamispiste: 1455 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011137
CAS-numero: 7440-02-0
UN: 3089
ADR: 4.1 II
Merck Index: 12,06582

Order Now

Testitulosten erittely

Appearance Black Powder
Total trace metal impurities ≤50 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR