Tris acetate 99%

Toimittaja: Thermo Fisher Scientific

434500250 434501000 434505000
ACRO434500250EA 68 EUR
ACRO434500250 ACRO434501000 ACRO434505000
Tris acetate 99%
Tris acetate
Kaava: NH₂C(CH₂OH)₃·CH₃COOH
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00038951
CAS-numero: 6850-28-8

Order Now

Testitulosten erittely

Appearance White Crystalline powder
Water ≤1.0 %
Solubility (1 g/ml in water) Clear colorless
UV at 290 nm A: ≤0.5
Titration with NaOH ≥98.5 %
Heavy metals (as Pb) ≤5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR