4-Aminopiperidine 95%

Toimittaja: Thermo Fisher Scientific
Vaara

Synonyms: Piperidin-4-amine

432920050 432920250
ACRO432920050EA 107 EUR
ACRO432920050 ACRO432920250
4-Aminopiperidine 95%
4-Aminopiperidine
Kaava: C₅H₁₂N₂
MW: 100.16 g/mol
Kiehumispiste: 65 °C (18Torr)
Tiheys: 0.954 g/cm³
Leimahduspiste: 46 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD02179399
CAS-numero: 13035-19-3
UN: 2733
ADR: 3 III

Order Now

Testitulosten erittely

Appearance Colorless to yellow to brown Liquid or low melting solid
Infrared spectrum Conforms
GC ≥94.0 %
UV at 260 nm A: ≤0.02
Titration after Ion exchange ≥99.0 % (on dry substance)
Membrane filtration test (on 0.5 M soln.) Passes test
Suitability (HPLC gradient) Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR