5-Amino-2-methylbenzothiazole 95%

Toimittaja: Thermo Fisher Scientific
Varoitus

432760010 432760050
ACRO432760010EA 42.1 EUR
ACRO432760010 ACRO432760050
5-Amino-2-methylbenzothiazole 95%
5-Amino-2-methylbenzothiazole
Kaava: C₈H₈N₂S
Sulamispiste: 101…103,5 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00226291
CAS-numero: 13382-43-9
UN: 2811
ADR: 6.1,III

Order Now

Testitulosten erittely

Appearance White to yellow Crystalline powder
Infrared spectrum Conforms
Melting point 100 to 105 °C
HPLC ≥94.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR