1,3-Benzothiazole-6-carboxaldehyde 97%

Toimittaja: Thermo Fisher Scientific
Varoitus

431150010 431150050
ACRO431150010EA 230 EUR
ACRO431150010 ACRO431150050
1,3-Benzothiazole-6-carboxaldehyde 97%
1,3-Benzothiazole-6-carboxaldehyde
Kaava: C₈H₅NOS
Sulamispiste: 92…93 °C
Storage Temperature: Jääkaappi
MDL numero: MFCD03086101
CAS-numero: 19989-67-4

Order Now

Testitulosten erittely

Appearance White to yellow Crystalline powder
Infrared spectrum Conforms
Melting point 90 to 96 °C
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR