6-Quinolineboronic acid 97%

Toimittaja: Thermo Fisher Scientific
Varoitus

430200010 430200050
ACRO430200010EA 146 EUR
ACRO430200010 ACRO430200050
6-Quinolineboronic acid 97%
6-Quinolineboronic acid
Kaava: C₉H₈BNO₂
Sulamispiste: 144…145 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD03093100
CAS-numero: 376581-24-7

Order Now

Testitulosten erittely

Appearance White to off-white Powder
Infrared spectrum Conforms
HPLC ≥96.0 %
GC ≥99.0 %
Color scale ≤25 APHA
Stabilizer Approx. 50 ppm BHT
Non-volatile matter ≤0.005 %
Acidity ≤0.0004 meq/g
Aluminium (Al) ≤0.5 ppm
Barium (Ba) ≤0.1 ppm
Calcium (Ca) ≤0.5 ppm
Cobalt (Co) ≤0.02 ppm
Copper (Cu) ≤0.02 ppm
Iron (Fe) ≤0.1 ppm
Lead (Pb) ≤0.1 ppm
Magnesium (Mg) ≤0.1 ppm
Manganese (Mn) ≤0.02 ppm
Nickel (Ni) ≤0.02 ppm
Tin (Sn) ≤0.1 ppm
Zinc (Zn) ≤0.1 ppm
Peroxides ≤0.005 % (as H2O2)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR