2-Chloro-3-formylpyridine 98%

Toimittaja: Thermo Fisher Scientific
Varoitus

429790010 429790050
ACRO429790010EA 39.7 EUR
ACRO429790010 ACRO429790050
2-Chloro-3-formylpyridine 98%
2-Chloro-3-formylpyridine
Kaava: C₆H₄ClNO
Sulamispiste: 47…50 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD01315308
CAS-numero: 36404-88-3
UN: 2811
ADR: 6.1,III

Order Now

Testitulosten erittely

Appearance Light yellow Crystalline powder or solid
Infrared spectrum Conforms
Melting point 47 to 52 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR