Diazolidinyl urea 95%

Toimittaja: Thermo Fisher Scientific
Varoitus

428580250 428582500
ACRO428580250EA 54 EUR
ACRO428580250 ACRO428582500
Diazolidinyl urea 95%
Diazolidinyl urea
Kaava: C₈H₁₄N₄O₇
Sulamispiste: 227…228 °C
Storage Temperature: Jääkaappi
MDL numero: MFCD03547942
CAS-numero: 78491-02-8

Order Now

Testitulosten erittely

Appearance White Powder
Infrared spectrum Conforms
Loss on drying ≤3 %
Solubility (50 mg/ml in water) Clear colorless
Elemental Analysis 19.0 to 21.5 % N
Hydroxyl value 102 to 124 mg KOH/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR