3-Methylisoquinoline 98%

Toimittaja: Thermo Fisher Scientific
Varoitus

428440010 428440050
ACRO428440010EA 78.2 EUR
ACRO428440010 ACRO428440050
3-Methylisoquinoline 98%
3-Methylisoquinoline
Kaava: C₁₀H₉N
MW: 143.19 g/mol
Kiehumispiste: 251 °C (760 mmHg)
Sulamispiste: 63…65 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00024139
CAS-numero: 1125-80-0
UN: 2811
ADR: 6.1 III

Order Now

Testitulosten erittely

Appearance White to yellow to brown Crystalline powder or solid
Infrared spectrum Authentic
Melting point 60 to 67 °C
HPLC ≥97.5 %
Density (15 °C) ≥0.79 kg/dm³
Aromatics ≤0.2 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR